Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190320.1 | NP_001177249.1 | 3 - 4 | 456 | 82 | |
NM_001190321.1 | NP_001177250.1 | 3 - 4 | 444 | 82 | ||
NM_020001.2 | NP_064385.1 | 4 - 5 | 546 | 82 | ||
GenBank mRNA | AF240357.1 | - | 4 - 5 | 515 | 82 | |
AF240358.1 | - | 3 - 4 | 413 | 82 | ||
AF240359.1 | - | 4 - 5 | 515 | 82 | ||
AK131697.1 | - | 4 - 5 | 533 | 82 | ||
AK154673.1 | - | 3 - 4 | 443 | 82 | ||
AK171046.1 | - | 5 - 6 | 716 | 82 | ||
BC023008.1 | - | 4 - 5 | 515 | 5376247 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |