Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700033757.mm.1 |
NM_020052.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033757.mm.1 |
NM_020052.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033757.mm.1 |
TC0700004084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033759.mm.1 |
NM_020052.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033759.mm.1 |
NM_020052.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033759.mm.1 |
TC0700004084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033756.mm.1 |
NM_020052.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033756.mm.1 |
NM_020052.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033756.mm.1 |
TC0700004084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033758.mm.1 |
NM_020052.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033758.mm.1 |
NM_020052.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033758.mm.1 |
TC0700004084.mm.1 |
MTA TC |