Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020577.2 | NP_065602.2 | 7 - 8 | 707 | 75 | |
GenBank mRNA | AF166383.1 | - | 7 - 8 | 677 | 75 | |
AK002801.1 | - | 7 - 8 | 640 | 75 | ||
AK006571.1 | - | 7 - 8 | 692 | 75 | ||
AK009814.1 | - | 6 - 7 | 672 | 75 | ||
AK011320.1 | - | 7 - 8 | 659 | 75 | ||
BC013468.1 | - | 7 - 8 | 632 | 75 | ||
BC145663.1 | - | 7 - 8 | 649 | 40130966 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |