Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF171060.1 | - | 3 - 4 | 226 | 91 | |
AK002399.1 | - | 3 - 4 | 247 | 91 | ||
AK171752.1 | - | 3 - 4 | 224 | 91 | ||
BC022715.1 | - | 3 - 4 | 212 | 91 | ||
BC037100.1 | - | 3 - 4 | 237 | 91 | ||
BC048821.1 | - | 3 - 4 | 234 | 3966642 | 91 | |
BG081026.2 | - | 3 - 4 | 193 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |