Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021339.2 | NP_067314.2 | 9 - 10 | 2044 | 71 | |
GenBank mRNA | AF090866.1 | - | 9 - 10 | 2044 | 71 | |
BC109176.2 | - | 9 - 10 | 2043 | 40048829 | 71 | |
CO810099.1 | - | 1 - 2 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |