Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301298.1 | NP_001288227.1 | 2 - 3 | 1280 | 66 | |
NM_021356.2 | NP_067331.2 | 2 - 3 | 1240 | 66 | ||
GenBank mRNA | AJ250669.1 | - | 2 - 3 | 387 | 66 | |
AK132176.1 | - | 2 - 3 | 1281 | 66 | ||
AK155423.1 | - | 2 - 3 | 1252 | 66 | ||
BC007483.1 | - | 2 - 3 | 1240 | 3495926 | 66 | |
BC094659.1 | - | 2 - 3 | 591 | 6854020 | 66 | |
CT010264.1 | - | 2 - 3 | 364 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |