Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021447.2 | NP_067422.1 | 4 - 5 | 903 | 82 | |
GenBank mRNA | AF294790.1 | - | 4 - 5 | 803 | 82 | |
BC107258.2 | - | 4 - 5 | 648 | 40052466 | 82 | |
BC107259.2 | - | 4 - 5 | 648 | 40052471 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |