Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021500.2 | NP_067475.2 | 7 - 8 | 904 | 62 | |
GenBank mRNA | AF263247.1 | - | 7 - 8 | 866 | 62 | |
AK034859.1 | - | 7 - 8 | 903 | 62 | ||
AK049424.1 | - | 7 - 8 | 924 | 62 | ||
AK075637.1 | - | 6 - 7 | 744 | 62 | ||
AK088854.1 | - | 7 - 8 | 912 | 62 | ||
AK145800.1 | - | 7 - 8 | 967 | 62 | ||
AK152523.1 | - | 7 - 8 | 912 | 62 | ||
AK168788.1 | - | 7 - 8 | 930 | 62 | ||
AK169149.1 | - | 7 - 8 | 912 | 62 | ||
BC039054.1 | - | 7 - 8 | 890 | 2648667 | 62 | |
BC058687.1 | - | 7 - 8 | 902 | 6411707 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |