Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10545893 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10545892 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10545894 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1418013_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1458044_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10545891 |
- |
- |
Mouse Genome 430 2.0 Array |
1418013_at |
- |
- |
Mouse Genome 430 2.0 Array |
1418013_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1458044_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1458044_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66116_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
66116_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0600020387.mm.1 |
TC0600002626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020387.mm.1 |
NM_023160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020387.mm.1 |
NM_023160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020386.mm.1 |
TC0600002626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020386.mm.1 |
NM_023160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020386.mm.1 |
NM_023160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020391.mm.1 |
TC0600002626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020391.mm.1 |
NM_023160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020391.mm.1 |
NM_023160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020384.mm.1 |
TC0600002626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020384.mm.1 |
NM_023160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020384.mm.1 |
NM_023160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020385.mm.1 |
TC0600002626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020385.mm.1 |
NM_023160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020385.mm.1 |
NM_023160.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020393.mm.1 |
TC0600002626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020393.mm.1 |
NM_023160.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020393.mm.1 |
NM_023160.2 |
Refseq |
Mouse Expression Set 430 |
1418013_at |
- |
- |
Mouse Expression Set 430 |
1418013_PM_at |
- |
- |
Mouse Expression Set 430 |
1458044_PM_at |
- |
- |
Mouse Expression Set 430 |
1458044_at |
- |
- |
Murine Genome U74v2 Set |
92835_at |
- |
- |
Murine Genome U74v2 Set |
92835_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418013_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1418013_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1418013_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1418013_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1458044_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1458044_at |
- |
- |