Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031187.4 | NP_112464.4 | 1 - 2 | 260 | 89 | |
NR_033521.1 | 1 - 2 | 260 | 89 | |||
GenBank mRNA | L00653.1 | - | 1 - 2 | 91 | 89 | |
U42405.1 | - | 1 - 2 | 77 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Detect Genomic DNA Mus musculus Probe spans exons |