Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200025028.mm.1 |
NM_001190817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025028.mm.1 |
NM_001190817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025028.mm.1 |
NM_007869.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025028.mm.1 |
NM_007869.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025028.mm.1 |
TC0200003033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025030.mm.1 |
NM_001190817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025030.mm.1 |
NM_001190817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025030.mm.1 |
NM_007869.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025030.mm.1 |
NM_007869.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025030.mm.1 |
TC0200003033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025027.mm.1 |
NM_001190817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025027.mm.1 |
NM_001190817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025027.mm.1 |
NM_007869.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025027.mm.1 |
NM_007869.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025027.mm.1 |
TC0200003033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200025031.mm.1 |
NM_001190817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025031.mm.1 |
NM_001190817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025031.mm.1 |
NM_007869.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025031.mm.1 |
NM_007869.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200025031.mm.1 |
TC0200003033.mm.1 |
MTA TC |