Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008185.3 | NP_032211.3 | 1 - 2 | 198 | 78 | |
GenBank mRNA | AK002338.1 | - | 1 - 2 | 198 | 78 | |
BC012254.1 | - | 1 - 2 | 113 | 4193816 | 78 | |
BC055020.1 | - | 1 - 2 | 134 | 4161144 | 78 | |
BY015234.1 | - | 1 - 2 | 185 | 78 | ||
W54467.1 | - | 1 - 2 | 149 | 78 | ||
X98055.1 | - | 1 - 2 | 110 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |