Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008318.3 | NP_032344.2 | 2 - 3 | 129 | 120 | |
GenBank mRNA | AK132371.1 | - | 2 - 3 | 128 | 120 | |
AK162640.1 | - | 2 - 3 | 112 | 120 | ||
BB618282.1 | - | 2 - 3 | 246 | 120 | ||
BC045143.1 | - | 2 - 3 | 114 | 6391115 | 120 | |
L20232.1 | - | 2 - 3 | 111 | 120 | ||
L23801.1 | - | 2 - 3 | 130 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |