Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008319.2 | NP_032345.2 | 6 - 7 | 1555 | 68 | |
GenBank mRNA | AK135040.1 | - | 6 - 7 | 1468 | 68 | |
BC113140.1 | - | 6 - 7 | 1503 | 40048010 | 68 | |
U06483.1 | - | 6 - 7 | 1503 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |