Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008324.2 | NP_032350.1 | 1 - 2 | 114 | 83 | |
GenBank mRNA | BC049931.1 | - | 1 - 2 | 105 | 5387937 | 83 |
M69109.1 | - | 1 - 2 | 171 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |