Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008378.2 | NP_032404.1 | 7 - 8 | 679 | 56 | |
GenBank mRNA | AK138136.1 | - | 7 - 8 | 650 | 56 | |
BC020524.1 | - | 7 - 8 | 655 | 4223620 | 56 | |
D87973.1 | - | 7 - 8 | 679 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |