Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008403.4 | NP_032429.1 | 2 - 3 | 258 | 81 | |
GenBank mRNA | AJ001373.1 | - | 2 - 3 | 240 | 81 | |
AK002786.1 | - | 2 - 3 | 215 | 81 | ||
AK004172.1 | - | 2 - 3 | 214 | 81 | ||
AK007635.1 | - | 2 - 3 | 216 | 81 | ||
AK075577.1 | - | 2 - 3 | 259 | 81 | ||
AK089937.1 | - | 2 - 3 | 372 | 81 | ||
AK147929.1 | - | 2 - 3 | 207 | 81 | ||
BC021151.1 | - | 2 - 3 | 397 | 81 | ||
BC028772.1 | - | 1 - 2 | 94 | 1230640 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |