Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008408.4 | NP_032434.3 | 4 - 5 | 338 | 64 | |
GenBank mRNA | AK030363.1 | - | 4 - 5 | 402 | 64 | |
AK077877.1 | - | 4 - 5 | 362 | 64 | ||
AK081547.1 | - | 4 - 5 | 394 | 64 | ||
AK129027.1 | - | 4 - 5 | 376 | 64 | ||
AK150640.1 | - | 4 - 5 | 383 | 64 | ||
AK151541.1 | - | 4 - 5 | 394 | 64 | ||
AK151791.1 | - | 4 - 5 | 394 | 64 | ||
AK152242.1 | - | 4 - 5 | 383 | 64 | ||
AK153840.1 | - | 4 - 5 | 339 | 64 | ||
AK160037.1 | - | 4 - 5 | 377 | 64 | ||
AK160607.1 | - | 4 - 5 | 339 | 64 | ||
AK168505.1 | - | 4 - 5 | 383 | 64 | ||
AK168937.1 | - | 4 - 5 | 383 | 64 | ||
AK169669.1 | - | 4 - 5 | 362 | 64 | ||
AX799090.1 | - | 4 - 5 | 381 | 64 | ||
BC028897.1 | - | 3 - 4 | 289 | 64 | ||
BC037612.1 | - | 4 - 5 | 377 | 5353797 | 64 | |
BC085313.1 | - | 4 - 5 | 367 | 30443880 | 64 | |
L34260.1 | - | 4 - 5 | 381 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |