Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008408.4 | NP_032434.3 | 6 - 7 | 573 | 115 | |
GenBank mRNA | AK030363.1 | - | 6 - 7 | 637 | 115 | |
AK077877.1 | - | 6 - 7 | 597 | 115 | ||
AK081547.1 | - | 6 - 7 | 629 | 115 | ||
AK129027.1 | - | 6 - 7 | 611 | 115 | ||
AK150640.1 | - | 6 - 7 | 618 | 115 | ||
AK151541.1 | - | 6 - 7 | 629 | 115 | ||
AK151791.1 | - | 6 - 7 | 629 | 115 | ||
AK152242.1 | - | 6 - 7 | 618 | 115 | ||
AK153840.1 | - | 6 - 7 | 574 | 115 | ||
AK153886.1 | - | 6 - 7 | 674 | 115 | ||
AK160037.1 | - | 6 - 7 | 612 | 115 | ||
AK160607.1 | - | 6 - 7 | 574 | 115 | ||
AK168505.1 | - | 6 - 7 | 618 | 115 | ||
AK168937.1 | - | 6 - 7 | 618 | 115 | ||
AK169669.1 | - | 6 - 7 | 597 | 115 | ||
AK206072.1 | - | 2 - 3 | 114 | 115 | ||
AK207675.1 | - | 2 - 3 | 114 | 115 | ||
AX799090.1 | - | 6 - 7 | 616 | 115 | ||
BC028897.1 | - | 5 - 6 | 524 | 115 | ||
BC037612.1 | - | 6 - 7 | 612 | 5353797 | 115 | |
BC085313.1 | - | 6 - 7 | 602 | 30443880 | 115 | |
L34260.1 | - | 6 - 7 | 616 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |