Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 23 - 24 | 3191 | 60 | |
GenBank mRNA | AK135430.1 | - | 23 - 24 | 2685 | 60 | |
AK136346.1 | - | 4 - 5 | 432 | 60 | ||
AK171938.1 | - | 23 - 24 | 3103 | 60 | ||
BC050946.1 | - | 23 - 24 | 3129 | 6822283 | 60 | |
BC054537.1 | - | 23 - 24 | 3113 | 60 | ||
D12646.1 | - | 23 - 24 | 3518 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |