Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008451.2 | NP_032477.2 | 6 - 7 | 1139 | 69 | |
NR_045528.1 | 6 - 7 | 1176 | 69 | |||
GenBank mRNA | AF055666.1 | - | 6 - 7 | 972 | 69 | |
AK141439.1 | - | 6 - 7 | 1069 | 69 | ||
AK185925.1 | - | 1 - 2 | 34 | 69 | ||
BC014845.1 | - | 6 - 7 | 983 | 2599082 | 69 | |
BM963360.1 | - | 2 - 3 | 310 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |