Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008453.5 | NP_032479.1 | 2 - 3 | 365 | 72 | |
GenBank mRNA | AK007959.1 | - | 2 - 3 | 364 | 72 | |
AK010713.1 | - | 2 - 3 | 310 | 72 | ||
AK046659.1 | - | 1 - 2 | 981 | 72 | ||
AK143838.1 | - | 2 - 3 | 368 | 72 | ||
AK157576.1 | - | 2 - 3 | 500 | 72 | ||
BC116938.1 | - | 2 - 3 | 332 | 40126257 | 72 | |
BC119214.1 | - | 2 - 3 | 332 | 8733963 | 72 | |
DQ981866.1 | - | 2 - 3 | 260 | 72 | ||
U36340.1 | - | 1 - 2 | 101 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |