Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008532.2 | NP_032558.2 | 2 - 3 | 524 | 69 | |
GenBank mRNA | AK145752.1 | - | 1 - 2 | 100 | 69 | |
AK167182.1 | - | 1 - 2 | 169 | 69 | ||
BC005618.1 | - | 1 - 2 | 165 | 3711069 | 69 | |
BC094465.1 | - | 1 - 2 | 190 | 4948935 | 69 | |
M76124.1 | - | 1 - 2 | 176 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |