Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009331.3 | NP_033357.1 | 7 - 8 | 718 | 59 | |
GenBank mRNA | AK037616.1 | - | 7 - 8 | 719 | 59 | |
AK038140.1 | - | 1 - 2 | 56 | 59 | ||
BC056630.1 | - | 6 - 7 | 960 | 59 | ||
BC138596.1 | - | 6 - 7 | 698 | 8861616 | 59 | |
BC145343.1 | - | 6 - 7 | 698 | 9053899 | 59 | |
X61385.1 | - | 8 - 9 | 860 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |