Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166584.1 | NP_001160056.1 | 9 - 10 | 1110 | 112 | |
NM_001166585.1 | NP_001160057.1 | 6 - 7 | 628 | 112 | ||
NM_009346.3 | NP_033372.1 | 8 - 9 | 1047 | 112 | ||
GenBank mRNA | AK140307.1 | - | 9 - 10 | 1111 | 112 | |
AK148375.1 | - | 8 - 9 | 1013 | 112 | ||
BC052021.1 | - | 6 - 7 | 569 | 112 | ||
BC060138.1 | - | 6 - 7 | 569 | 6410573 | 112 | |
L06865.1 | - | 6 - 7 | 574 | 112 | ||
L13853.1 | - | 8 - 9 | 774 | 112 | ||
S74227.1 | - | 7 - 8 | 1035 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |