Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB000619.1 | - | 2 - 3 | 281 | 117 | |
BC141033.1 | - | 1 - 2 | 175 | 9055645 | 117 | |
BC145184.1 | - | 1 - 2 | 175 | 117 | ||
BC145185.1 | - | 1 - 2 | 175 | 9053736 | 117 | |
BC145186.1 | - | 1 - 2 | 175 | 9053737 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |