Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293561.1 | NP_001280490.1 | 13 - 14 | 2344 | 65 | |
NM_009472.4 | NP_033498.1 | 12 - 13 | 2287 | 65 | ||
GenBank mRNA | AK031655.1 | - | 13 - 14 | 2311 | 65 | |
AK138987.1 | - | 1 - 2 | 84 | 65 | ||
AK141181.1 | - | 13 - 14 | 2310 | 65 | ||
AK142411.1 | - | 13 - 14 | 2310 | 65 | ||
AK142974.1 | - | 13 - 14 | 2238 | 65 | ||
AK164239.1 | - | 12 - 13 | 2088 | 65 | ||
BC115772.1 | - | 13 - 14 | 2334 | 40085998 | 65 | |
U72634.1 | - | 12 - 13 | 2287 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |