Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293561.1 | NP_001280490.1 | 15 - 16 | 2657 | 68 | |
NM_009472.4 | NP_033498.1 | 14 - 15 | 2600 | 68 | ||
GenBank mRNA | AK031655.1 | - | 15 - 16 | 2624 | 68 | |
AK138987.1 | - | 3 - 4 | 397 | 68 | ||
AK141181.1 | - | 15 - 16 | 2623 | 68 | ||
AK142411.1 | - | 15 - 16 | 2623 | 68 | ||
AK142974.1 | - | 15 - 16 | 2551 | 68 | ||
AK164239.1 | - | 14 - 15 | 2401 | 68 | ||
BC115772.1 | - | 15 - 16 | 2647 | 40085998 | 68 | |
U72634.1 | - | 14 - 15 | 2600 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |