Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF324471.1 | - | 3 - 4 | 2099 | 106 | |
AK142052.1 | - | 7 - 8 | 2552 | 106 | ||
AK142210.1 | - | 8 - 9 | 2589 | 106 | ||
BC141284.1 | - | 7 - 8 | 2402 | 9055896 | 106 | |
BC145612.1 | - | 7 - 8 | 2402 | 9054178 | 106 | |
X95503.1 | - | 9 - 10 | 2506 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |