Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009541.2 | NP_033567.2 | 6 - 7 | 880 | 91 | |
GenBank mRNA | AK051971.1 | - | 6 - 7 | 870 | 91 | |
AK134251.1 | - | 6 - 7 | 840 | 91 | ||
BC060055.1 | - | 6 - 7 | 854 | 91 | ||
BC145312.1 | - | 5 - 6 | 753 | 9053867 | 91 | |
BC145313.1 | - | 5 - 6 | 753 | 91 | ||
BC145821.1 | - | 5 - 6 | 753 | 40131124 | 91 | |
U14556.1 | - | 6 - 7 | 878 | 91 | ||
U22396.1 | - | 6 - 7 | 868 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |