Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009568.3 | NP_033594.1 | 3 - 4 | 326 | 59 | |
GenBank mRNA | AK017108.1 | - | 3 - 4 | 285 | 59 | |
BC005437.1 | - | 3 - 4 | 326 | 3591835 | 59 | |
BC095945.1 | - | 2 - 3 | 146 | 4936327 | 59 | |
CJ174209.1 | - | 3 - 4 | 228 | 59 | ||
U46187.1 | - | 3 - 4 | 269 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |