Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010072.3 | NP_034202.1 | 1 - 2 | 189 | 73 | |
GenBank mRNA | AB004789.1 | - | 1 - 2 | 163 | 73 | |
AI099209.1 | - | 1 - 2 | 189 | 73 | ||
AK004834.1 | - | 1 - 2 | 182 | 73 | ||
AK005268.1 | - | 1 - 2 | 175 | 73 | ||
AK008548.1 | - | 1 - 2 | 177 | 73 | ||
BC061151.1 | - | 1 - 2 | 169 | 30307514 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |