Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010296.2 | NP_034426.2 | 3 - 4 | 254 | 101 | |
GenBank mRNA | AB025922.1 | - | 3 - 4 | 264 | 101 | |
AB234616.1 | - | 4 - 5 | 765 | 101 | ||
AB234617.1 | - | 3 - 4 | 646 | 101 | ||
AF026305.1 | - | 3 - 4 | 254 | 101 | ||
BC131649.1 | - | 2 - 3 | 232 | 101 | ||
BC131650.1 | - | 2 - 3 | 232 | 40110429 | 101 | |
CJ062804.1 | - | 2 - 3 | 238 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |