Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010357.3 | NP_034487.2 | 5 - 6 | 528 | 69 | |
GenBank mRNA | AI043064.1 | - | 2 - 3 | 69 | ||
AK008189.1 | - | 5 - 6 | 514 | 69 | ||
AK008193.1 | - | 6 - 7 | 479 | 69 | ||
AK008400.1 | - | 5 - 6 | 512 | 69 | ||
AK008490.1 | - | 5 - 6 | 479 | 69 | ||
AK009668.1 | - | 5 - 6 | 482 | 69 | ||
AK010098.1 | - | 5 - 6 | 482 | 69 | ||
AK011177.1 | - | 5 - 6 | 479 | 69 | ||
AK011841.1 | - | 5 - 6 | 507 | 69 | ||
AK019271.1 | - | 5 - 6 | 503 | 69 | ||
BC012639.1 | - | 5 - 6 | 482 | 3995378 | 69 | |
BU953110.1 | - | 5 - 6 | 531 | 69 | ||
L06047.1 | - | 5 - 6 | 480 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |