Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080746.2 | NP_001074215.1 | 33 - 34 | 3319 | 62 | |
NM_001080747.2 | NP_001074216.1 | 32 - 33 | 3256 | 62 | ||
NM_001080748.2 | NP_001074217.1 | 31 - 32 | 3199 | 62 | ||
NM_001080749.2 | NP_001074218.1 | 31 - 32 | 3211 | 62 | ||
NM_010365.4 | NP_034495.2 | 32 - 33 | 3262 | 62 | ||
GenBank mRNA | AF017085.1 | - | 33 - 34 | 2963 | 62 | |
AF043219.1 | - | 31 - 32 | 2883 | 62 | ||
AF043220.1 | - | 30 - 31 | 2820 | 62 | ||
AK013348.1 | - | 10 - 11 | 796 | 62 | ||
AK145698.1 | - | 23 - 24 | 2147 | 62 | ||
AK147201.1 | - | 31 - 32 | 3169 | 62 | ||
AK183312.1 | - | 4 - 5 | 139 | 62 | ||
AK211276.1 | - | 4 - 5 | 139 | 62 | ||
AY030290.1 | - | 32 - 33 | 2949 | 62 | ||
AY030291.1 | - | 32 - 33 | 2955 | 62 | ||
AY030292.1 | - | 31 - 32 | 2892 | 62 | ||
AY030293.1 | - | 33 - 34 | 3012 | 62 | ||
BC047387.1 | - | 31 - 32 | 2858 | 62 | ||
BC053044.1 | - | 33 - 34 | 3288 | 5704935 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |