Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136067.2 | NP_001129539.1 | 8 - 9 | 1154 | 82 | |
NM_001161421.1 | NP_001154893.1 | 8 - 9 | 1151 | 82 | ||
NM_001161422.1 | NP_001154894.1 | 7 - 8 | 972 | 82 | ||
NM_001161423.1 | NP_001154895.1 | 7 - 8 | 1024 | 82 | ||
NM_001161424.1 | NP_001154896.1 | 7 - 8 | 1021 | 82 | ||
NM_010547.2 | NP_034677.2 | 7 - 8 | 1050 | 82 | ||
NM_178590.4 | NP_848705.1 | 7 - 8 | 1047 | 82 | ||
GenBank mRNA | AF069542.1 | - | 6 - 7 | 909 | 82 | |
AF513109.1 | - | 8 - 9 | 1065 | 82 | ||
AK037020.1 | - | 7 - 8 | 1026 | 82 | ||
AK154095.1 | - | 7 - 8 | 1036 | 82 | ||
AY112937.1 | - | 8 - 9 | 1154 | 82 | ||
BC021431.1 | - | 8 - 9 | 1141 | 5030381 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |