Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038607.2 | NP_001033696.1 | 1 - 2 | 312 | 77 | |
NM_010600.3 | NP_034730.1 | 1 - 2 | 312 | 77 | ||
GenBank mRNA | AK140193.1 | - | 1 - 2 | 218 | 77 | |
AK147461.1 | - | 1 - 2 | 306 | 77 | ||
BC109012.1 | - | 1 - 2 | 171 | 40048708 | 77 | |
BC109013.1 | - | 1 - 2 | 171 | 40048709 | 77 | |
CJ104826.1 | - | 1 - 2 | 315 | 77 | ||
U04294.1 | - | 1 - 2 | 194 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |