Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083188.1 | NP_001076657.1 | 19 - 20 | 2078 | 93 | |
NM_001199310.1 | NP_001186239.1 | 19 - 20 | 2086 | 93 | ||
NM_010715.2 | NP_034845.2 | 19 - 20 | 2003 | 93 | ||
GenBank mRNA | AK153993.1 | - | 19 - 20 | 1965 | 93 | |
AK160208.1 | - | 4 - 5 | 372 | 93 | ||
AK161793.1 | - | 12 - 13 | 1161 | 93 | ||
AK168643.1 | - | 19 - 20 | 2082 | 93 | ||
BC028287.1 | - | 19 - 20 | 1928 | 4911261 | 93 | |
BC138541.1 | - | 19 - 20 | 2086 | 8861561 | 93 | |
BC138542.1 | - | 19 - 20 | 2086 | 8861562 | 93 | |
U04674.1 | - | 18 - 19 | 1871 | 93 | ||
U19604.1 | - | 19 - 20 | 2116 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |