Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040435.2 | NP_001035525.1 | 12 - 13 | 2174 | 112 | |
GenBank mRNA | AF093542.1 | - | 13 - 14 | 1408 | 112 | |
AF156934.1 | - | 12 - 13 | 1728 | 112 | ||
AF203445.1 | - | 12 - 13 | 1675 | 112 | ||
AF203446.1 | - | 12 - 13 | 1063 | 112 | ||
AF247674.1 | - | 13 - 14 | 1459 | 112 | ||
AK145556.1 | - | 12 - 13 | 1735 | 112 | ||
AK160291.1 | - | 12 - 13 | 1737 | 112 | ||
BC003252.1 | - | 12 - 13 | 1731 | 3500413 | 112 | |
BC055071.1 | - | 4150 | 112 | |||
BC066184.1 | - | 12 - 13 | 1710 | 30457053 | 112 | |
BC068314.1 | - | 12 - 13 | 1710 | 30461783 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |