Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136259.1 | NP_001129731.1 | 3 - 4 | 306 | 92 | |
NM_011622.3 | NP_035752.1 | 3 - 4 | 306 | 92 | ||
GenBank mRNA | AJ006972.1 | - | 3 - 4 | 283 | 92 | |
AK028398.1 | - | 3 - 4 | 247 | 92 | ||
AK051245.1 | - | 3 - 4 | 288 | 92 | ||
AK150143.1 | - | 3 - 4 | 256 | 92 | ||
AK160403.1 | - | 3 - 4 | 252 | 92 | ||
AK184431.1 | - | 3 - 4 | 182 | 92 | ||
AK201506.1 | - | 2 - 3 | 182 | 92 | ||
BC021633.1 | - | 3 - 4 | 269 | 4208226 | 92 | |
BC093520.1 | - | 3 - 4 | 234 | 2866013 | 92 | |
BC157905.1 | - | 3 - 4 | 267 | 9088106 | 92 | |
BC158042.1 | - | 3 - 4 | 267 | 9007343 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |