Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011707.2 | NP_035837.1 | 2 - 3 | 412 | 75 | |
GenBank mRNA | AF440693.1 | - | 2 - 3 | 182 | 75 | |
AK011736.1 | - | 2 - 3 | 267 | 75 | ||
AK090325.1 | - | 2 - 3 | 267 | 75 | ||
AK167494.1 | - | 2 - 3 | 267 | 75 | ||
BC012690.1 | - | 2 - 3 | 258 | 4193767 | 75 | |
CK617609.1 | - | 2 - 3 | 475 | 75 | ||
M77123.1 | - | 2 - 3 | 182 | 75 | ||
X63003.1 | - | 2 - 3 | 244 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |