Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011707.2 | NP_035837.1 | 3 - 4 | 758 | 76 | |
GenBank mRNA | AK011736.1 | - | 3 - 4 | 613 | 76 | |
AK090325.1 | - | 3 - 4 | 613 | 76 | ||
AK167494.1 | - | 3 - 4 | 611 | 76 | ||
M77123.1 | - | 3 - 4 | 528 | 76 | ||
X63003.1 | - | 3 - 4 | 590 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |