Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xirp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011724.3 | NP_035854.3 | 1 - 2 | 165 | 61 | |
GenBank mRNA | AF051945.2 | - | 1 - 2 | 136 | 61 | |
BC117959.1 | - | 1 - 2 | 70 | 40098186 | 61 | |
BC117960.1 | - | 1 - 2 | 70 | 40098187 | 61 | |
BY783852.1 | - | 1 - 2 | 165 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |