Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040692.3 | NP_001035782.1 | 2 - 3 | 398 | 65 | |
NM_001136087.2 | NP_001129559.1 | 2 - 3 | 398 | 65 | ||
NM_001168643.1 | NP_001162114.1 | 2 - 3 | 398 | 65 | ||
NM_001168644.1 | NP_001162115.1 | 2 - 3 | 398 | 65 | ||
NM_001168645.1 | NP_001162116.1 | 2 - 3 | 398 | 65 | ||
NM_001168646.1 | NP_001162117.1 | 2 - 3 | 398 | 65 | ||
GenBank mRNA | AF075262.1 | - | 2 - 3 | 375 | 65 | |
AF075263.1 | - | 2 - 3 | 375 | 65 | ||
AF075264.1 | - | 2 - 3 | 375 | 65 | ||
AF075265.1 | - | 2 - 3 | 375 | 65 | ||
AF075266.1 | - | 2 - 3 | 375 | 65 | ||
AF075267.1 | - | 2 - 3 | 375 | 65 | ||
AK085258.1 | - | 2 - 3 | 395 | 65 | ||
AK085473.1 | - | 2 - 3 | 396 | 65 | ||
AK128907.1 | - | 2 - 3 | 395 | 65 | ||
BC010748.1 | - | 2 - 3 | 374 | 4235659 | 65 | |
CA491609.1 | - | 2 - 3 | 435 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |