Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK047537.1 | - | 9 - 10 | 1194 | 100 | |
AK089383.1 | - | 9 - 10 | 1158 | 100 | ||
AK171139.1 | - | 9 - 10 | 1169 | 100 | ||
AK210877.1 | - | 2 - 3 | 108 | 101 | ||
BC053416.1 | - | 8 - 9 | 1018 | 6533681 | 100 | |
U00978.1 | - | 8 - 9 | 913 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |