Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012033.3 | NP_036163.3 | 9 - 10 | 1323 | 93 | |
GenBank mRNA | AF153366.1 | - | 9 - 10 | 1265 | 93 | |
BC010745.1 | - | 9 - 10 | 1268 | 4239192 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |