Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013711.3 | NP_038739.2 | 15 - 16 | 1390 | 65 | |
GenBank mRNA | AB027566.1 | - | 15 - 16 | 1390 | 65 | |
AF136399.1 | - | 15 - 16 | 1393 | 65 | ||
AF171053.1 | - | 15 - 16 | 1356 | 65 | ||
AF412308.1 | - | 14 - 15 | 1299 | 65 | ||
AK075828.1 | - | 15 - 16 | 1368 | 65 | ||
BC029344.1 | - | 15 - 16 | 1370 | 65 | ||
BC052157.1 | - | 15 - 16 | 1369 | 6506286 | 65 | |
BC052758.1 | - | 15 - 16 | 1369 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |