Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013806.2 | NP_038834.2 | 25 - 26 | 3707 | 69 | |
GenBank mRNA | AF227274.1 | - | 25 - 26 | 3611 | 69 | |
AF282772.1 | - | 25 - 26 | 3611 | 69 | ||
AF282773.1 | - | 25 - 26 | 3611 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |