Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013841.3 | NP_038869.1 | 11 - 12 | 1951 | 69 | |
GenBank mRNA | AK146190.1 | - | 11 - 12 | 1353 | 69 | |
AK168101.1 | - | 11 - 12 | 1353 | 69 | ||
BC012691.1 | - | 11 - 12 | 1327 | 4038142 | 69 | |
BC058528.1 | - | 11 - 12 | 1343 | 5718352 | 69 | |
U66865.1 | - | 11 - 12 | 1966 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |