Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077595.2 | NP_001071063.1 | 4 - 5 | 3604 | 109 | |
NM_001077596.2 | NP_001071064.1 | 4 - 5 | 3652 | 109 | ||
NM_015756.2 | NP_056571.2 | 5 - 6 | 3976 | 109 | ||
GenBank mRNA | AF199421.1 | - | 5 - 6 | 3976 | 109 | |
AF199422.1 | - | 4 - 5 | 3544 | 109 | ||
AK049347.1 | - | 1 - 2 | 1510 | 109 | ||
AK129371.1 | - | 1 - 2 | 2982 | 109 | ||
AK147493.1 | - | 4 - 5 | 3587 | 109 | ||
AK164918.1 | - | 4 - 5 | 3563 | 109 | ||
BC145113.1 | - | 6 - 7 | 3393 | 9053661 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |